ContactUsLinkComponent
Human PD-1, Fc,His Tag (PD1-H5256) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1 ). 1 * 100 µG
Catalog # ACRBPD1-H5256100UG
Supplier:  ACRO Biosystems
Human PD-1, Fc,His Tag (PD1-H5256) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1 ). 1 * 100 µG
Catalog # ACRBPD1-H5256100UG
Supplier:  ACRO Biosystems
Frequently Bought Together