ContactUsLinkComponent
Human BTLA (31-150), His Tag (BTA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AA P44003.1). 1 * 50 µG
Katalognr. ACRBBTA-H52H3-50UG
Supplier:  ACRO Biosystems
Human BTLA (31-150), His Tag (BTA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AA P44003.1). 1 * 50 µG
Katalognr. ACRBBTA-H52H3-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together