ContactUsLinkComponent
Human TRBC2, His Tag (TR2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 1 - Ala 144 (Accession # A0A5B9-1). 1 * 100 µG
Catalog # ACRBTR2-H52H3100UG
Supplier:  ACRO Biosystems
Human TRBC2, His Tag (TR2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 1 - Ala 144 (Accession # A0A5B9-1). 1 * 100 µG
Catalog # ACRBTR2-H52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together