ContactUsLinkComponent
Mouse GFAP, His Tag (GFP-M5148) is expressed from E.coli cells. It contains AA Met 1 - Met 430 (Accession # P03995-1). 1 * 100 µG
Catalog # ACRBGFP-M5148100UG
Supplier:  ACRO Biosystems
Mouse GFAP, His Tag (GFP-M5148) is expressed from E.coli cells. It contains AA Met 1 - Met 430 (Accession # P03995-1). 1 * 100 µG
Catalog # ACRBGFP-M5148100UG
Supplier:  ACRO Biosystems
Frequently Bought Together