ContactUsLinkComponent
PE-Labeled Human FAP, His Tag (FAP-HP245) is produced via site-specific conjugation of PE to Human FAP, His Tag under optimal condit ions with a proprietary technology. Human FAP, His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 1 * 200 Tests
Catalog # ACRBFAP-HP245-200
Supplier:  ACRO Biosystems
PE-Labeled Human FAP, His Tag (FAP-HP245) is produced via site-specific conjugation of PE to Human FAP, His Tag under optimal condit ions with a proprietary technology. Human FAP, His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 1 * 200 Tests
Catalog # ACRBFAP-HP245-200
Supplier:  ACRO Biosystems
Frequently Bought Together