ContactUsLinkComponent
SARS-CoV-2 Spike S2 protein, His Tag (BA.2/Omicron) (S2N-C52Hh) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1 (N764K, D796Y, Q954H, N969K, F817P, A892P, A899P, A942P, K986P, V987P)). The spike d 1 * 1 mg
Catalog # ACRBS2N-C52HH-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 Spike S2 protein, His Tag (BA.2/Omicron) (S2N-C52Hh) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1 (N764K, D796Y, Q954H, N969K, F817P, A892P, A899P, A942P, K986P, V987P)). The spike d 1 * 1 mg
Catalog # ACRBS2N-C52HH-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together