ContactUsLinkComponent
SARS-CoV-2 Spike Trimer, His Tag (BA.5/Omicron) (SPN-C522e) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, HV69-70del, G142D, V213G, G339D, S371F, S373P, S375F, T376A, D4 1 * 50 µG
Catalog # ACRBSPN-C522E-50UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Spike Trimer, His Tag (BA.5/Omicron) (SPN-C522e) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, HV69-70del, G142D, V213G, G339D, S371F, S373P, S375F, T376A, D4 1 * 50 µG
Catalog # ACRBSPN-C522E-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together