ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid protein, His Tag (BA.5/Omicron) (NUN-C52Hx) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1 (P13L, ERS31-33del, E136D, R203K, G204R, S413R)). The mutations are identified on O 1 * 100 µG
Catalog # ACRBNUN-C52HX100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid protein, His Tag (BA.5/Omicron) (NUN-C52Hx) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1 (P13L, ERS31-33del, E136D, R203K, G204R, S413R)). The mutations are identified on O 1 * 100 µG
Catalog # ACRBNUN-C52HX100UG
Supplier:  ACRO Biosystems
Frequently Bought Together