ContactUsLinkComponent
SFTS virus Gn Protein, His Tag (GNN-S52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Lys 452 (Accession # F1BA47-1). 1 * 500 µG
Catalog # ACRBGNN-S52H3500UG
Supplier:  ACRO Biosystems
SFTS virus Gn Protein, His Tag (GNN-S52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Lys 452 (Accession # F1BA47-1). 1 * 500 µG
Catalog # ACRBGNN-S52H3500UG
Supplier:  ACRO Biosystems
Frequently Bought Together