ContactUsLinkComponent
Influenza A [A/Bangkok/1/1979 (H3N2)] HA, His Tag (HA2-V52H3) is expressed from human 293 cells (HEK293). It contains AA Asn 18 - Tr p 530 (Accession # AFM68976.1). 1 * 100 µG
Catalog # ACRBHA2-V52H3100UG
Supplier:  ACRO Biosystems
Influenza A [A/Bangkok/1/1979 (H3N2)] HA, His Tag (HA2-V52H3) is expressed from human 293 cells (HEK293). It contains AA Asn 18 - Tr p 530 (Accession # AFM68976.1). 1 * 100 µG
Catalog # ACRBHA2-V52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together