ContactUsLinkComponent
SARS-CoV-2 Spike RBD, His Tag (BA.2.12.1/Omicron) (SPD-C522q) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - L ys 537 (Accession # QHD43416.1 (G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, L452Q, S477N, T478K, E 1 * 100 µG
Catalog # ACRBSPD-C522Q100UG
Anbieter:  ACROBIOSYSTEMS
SARS-CoV-2 Spike RBD, His Tag (BA.2.12.1/Omicron) (SPD-C522q) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - L ys 537 (Accession # QHD43416.1 (G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, L452Q, S477N, T478K, E 1 * 100 µG
Catalog # ACRBSPD-C522Q100UG
Anbieter:  ACROBIOSYSTEMS
Frequently Bought Together