ContactUsLinkComponent
SARS-CoV-2 Spike RBD, His Tag (BA.2.13/Omicron) (SPD-C522p) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, L452M, S477N, T478K, E48 1 * 100 µG
Catalog # ACRBSPD-C522P100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Spike RBD, His Tag (BA.2.13/Omicron) (SPD-C522p) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, L452M, S477N, T478K, E48 1 * 100 µG
Catalog # ACRBSPD-C522P100UG
Supplier:  ACRO Biosystems
Frequently Bought Together