ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike RBD (R346S, N394S, Y449N, E484K, F490S, N501Y), His,Avitag (SPD-C82Er) is expressed from human 293 cel ls (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (R346S, N394S, Y449N, E484K, F490S, N501Y)). Th 1 * 200 µG
Catalog # ACRBSPD-C82ER200UG
Anbieter:  ACROBIOSYSTEMS
Biotinylated SARS-CoV-2 Spike RBD (R346S, N394S, Y449N, E484K, F490S, N501Y), His,Avitag (SPD-C82Er) is expressed from human 293 cel ls (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (R346S, N394S, Y449N, E484K, F490S, N501Y)). Th 1 * 200 µG
Catalog # ACRBSPD-C82ER200UG
Anbieter:  ACROBIOSYSTEMS
Frequently Bought Together