ContactUsLinkComponent
SARS-CoV-2 Spike RBD, His Tag (BA.2/Omicron) (SPD-C522g) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 53 7 (Accession # QHD43416.1 (G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, S477N, T478K, E484A, Q493R, 1 * 100 µG
  ACRBSPD-C522G100UG
SARS-CoV-2 Spike RBD, His Tag (BA.2/Omicron) (SPD-C522g) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 53 7 (Accession # QHD43416.1 (G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, S477N, T478K, E484A, Q493R, 1 * 100 µG
  ACRBSPD-C522G100UG
Frequently Bought Together