Human Fc gamma RIIIA / CD16a (F176, S197P), His Tag (CDA-H52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (F176, S197P) (Accession # P08637-1 (F176, S197P)). 1 * 1 mg
Katalognr. ACRBCDA-H52H6-1MG
Supplier: ACRO Biosystems
Frequently Bought Together