ContactUsLinkComponent
Biotinylated Human Siglec-8, His,Avitag (SI8-H82E9) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Acc ession # Q9NYZ4-1). 1 * 25 µG
Catalog # ACRBSI8-H82E9-25UG
Supplier:  ACRO Biosystems
Biotinylated Human Siglec-8, His,Avitag (SI8-H82E9) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Acc ession # Q9NYZ4-1). 1 * 25 µG
Catalog # ACRBSI8-H82E9-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together