ContactUsLinkComponent
Human EDIL3, Fc Tag (ED3-H5259) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Glu 480 (Accession # O43854-1). 1 * 1 mg
Catalog # ACRBED3-H5259-1MG
Supplier:  ACRO Biosystems
Human EDIL3, Fc Tag (ED3-H5259) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Glu 480 (Accession # O43854-1). 1 * 1 mg
Catalog # ACRBED3-H5259-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together