ContactUsLinkComponent
Biotinylated Cynomolgus Serum Albumin, His,Avitag (CSA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Al a 608 (Accession # A2V9Z4-1). 1 * 200 µG
Catalog # ACRBCSA-C82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus Serum Albumin, His,Avitag (CSA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Al a 608 (Accession # A2V9Z4-1). 1 * 200 µG
Catalog # ACRBCSA-C82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together