ContactUsLinkComponent
Human LRRC32&TGFB1 Heterotrimer protein, His Tag&Tag Free (GA1-H52W9) is expressed from human 293 cells (HEK293). It contains AA His 20 - Asn 627 (LRRC32) & Leu 30 - Ser 390 (TGF-beta 1) (Accession # Q14392-1 (LRRC32) & P01137-1 (TGF-beta1)). 1 * 100 µG
Catalog # ACRBGA1-H52W9100UG
Supplier:  ACRO Biosystems
Human LRRC32&TGFB1 Heterotrimer protein, His Tag&Tag Free (GA1-H52W9) is expressed from human 293 cells (HEK293). It contains AA His 20 - Asn 627 (LRRC32) & Leu 30 - Ser 390 (TGF-beta 1) (Accession # Q14392-1 (LRRC32) & P01137-1 (TGF-beta1)). 1 * 100 µG
Catalog # ACRBGA1-H52W9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together