ContactUsLinkComponent
Human CD43, His Tag (CD3-H52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Arg 253 (Accession # P16150-1 ). 1 * 100 µG
Catalog # ACRBCD3-H52H9100UG
Supplier:  ACRO Biosystems
Human CD43, His Tag (CD3-H52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Arg 253 (Accession # P16150-1 ). 1 * 100 µG
Catalog # ACRBCD3-H52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together