ContactUsLinkComponent
Biotinylated Human Complement C3, His,Avitag (CO3-H82E8) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Asn 166 3 (Accession # P01024-1). 1 * 200 µG
Catalog # ACRBCO3-H82E8200UG
Supplier:  ACRO Biosystems
Biotinylated Human Complement C3, His,Avitag (CO3-H82E8) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Asn 166 3 (Accession # P01024-1). 1 * 200 µG
Catalog # ACRBCO3-H82E8200UG
Supplier:  ACRO Biosystems
Frequently Bought Together