ContactUsLinkComponent
Cynomolgus Fc epsilon RI alpha, His Tag (FCA-C52H9) is expressed from human 293 cells (HEK293). It contains AA Val 29 - Lys 204 (Acc ession # A0A7N9DA09-1). 1 * 1 mg
Catalog # ACRBFCA-C52H9-1MG
Supplier:  ACRO Biosystems
Cynomolgus Fc epsilon RI alpha, His Tag (FCA-C52H9) is expressed from human 293 cells (HEK293). It contains AA Val 29 - Lys 204 (Acc ession # A0A7N9DA09-1). 1 * 1 mg
Catalog # ACRBFCA-C52H9-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together