ContactUsLinkComponent
Human IL-23A&IL-12B Heterodimer Protein (ILB-H5219) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 (IL- 23A) & Arg 20 - Pro 189 (IL-12B) (Accession # Q9NPF7-1 (IL23A) & P29460-1 (IL12B)). 1 * 1 mg
Catalog # ACRBILB-H5219-1MG
Supplier:  ACRO Biosystems
Human IL-23A&IL-12B Heterodimer Protein (ILB-H5219) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Ser 328 (IL- 23A) & Arg 20 - Pro 189 (IL-12B) (Accession # Q9NPF7-1 (IL23A) & P29460-1 (IL12B)). 1 * 1 mg
Catalog # ACRBILB-H5219-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together