[EN]ANTI-FLAG MAGNETIC BEADS 1 * 1 mL
Catalog # MDEXHY-K0207-1ML
Supplier: MEDCHEMEXPRESS
Frequently Bought Together
Lugol's iodine solution, Sigma-Aldrich®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips, nitrite, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
Filling solutions for pH electrodes, Orion™
Supplier: Thermo Orion
Test strips, lactic acid, Reflectoquant®, Supelco®
Supplier: Merck
Joint grease, K.W.S.
Supplier: POLLATH
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Paramat, Solidification point: 55 - 58 °C, in pastille form, Gurr™, without DMSO
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond