ContactUsLinkComponent
Masterflex® Fitting, Polycarbonate, RQX Male Connector Plug; 25/PK
  MFLX40618-49
undefined
Masterflex® Fitting, Polycarbonate, RQX Male Connector Plug; 25/PK
  MFLX40618-49
Disconnect and reconnect your flow path with ease.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Large ergonomic button and thumb pad with secure latch and optional integrated locking mechanism
  • Several connector configurations available in commonly used polypropylene and polycarbonate materials
  • Plugs and caps available to maintain fluid path integrity
RQ Series offers a cost effective solution for flexible tubing connections in single use systems. The connector set features a user-friendly interface that includes a large guarded button for a secure lock to guard against disconnects. Use these fittings to quickly connect and disconnect from your flow line. Simply order a body (female half) and an insert (male half).
  • Connection Type:
    Quick-Disconnect Plug
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polycarbonate, RQX Male Connector Plug; 25/PK
  • Environmentally Preferable:
  • Warranty:
    Shelf life
  • Pk:
    1
  • Fitting material:
    Polycarbonate
Frequently Bought Together