Order Entry
Switzerland
ContactUsLinkComponent
Masterflex® Fitting, PVDF, Straight, Hose Barb Reducer, 1" ID x 1/2" ID; 10/PK
Masterflex® Fitting, PVDF, Straight, Hose Barb Reducer, 1" ID x 1/2" ID; 10/PK
Catalog # MFLX40614-78
undefined
Masterflex® Fitting, PVDF, Straight, Hose Barb Reducer, 1" ID x 1/2" ID; 10/PK
Catalog # MFLX40614-78
  • Model:
  • 30703-56
  • 30703-55
  • 40703-41
  • 30616-12
  • 30703-49
  • Orifice dimensions:
  • 8,73 mm (0,34375")
  • 3,175 mm (0,125")
  • 4,763 mm (0,1875")
  • 0,0625"
  • 0,03125"
  • 1,588 mm (0,0625")
  • 0,03125"
  • 15,08 mm (0,59375")
  • 0,03125"
  • 4,763 mm (0,1875")
  • 3,175 mm (0,125")
  • 0,7925 mm (0,03125")
  • 3,175 mm (0,125")
  • 4,763 mm (0,1875")
  • 4,763 mm (0,1875")
  • 3,175 mm (0,125")
  • 0,7925 mm (0,03125")
  • 6,35 mm (0,25")
  • 3,175 mm (0,125")
  • 6,35 mm (0,25")
  • 11,9 mm (0,46875")
  • 11,9 mm (0,46875")
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ETO
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Currently Viewing

Specifications

About this item

Best for low-pressure applications.

  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range −80 to 265 °F (−62 to 129 °C)

These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.

  • Barb size/tubing int.Ø (port 1):
    1"
  • Barb size/tubing int.Ø (port 2):
    ½"
  • Connection Type:
    Barbed Reducer
  • Flow pattern:
    Straight
  • Description:
    Fitting, PVDF, Straight, Hose Barb Reducer, 1" ID x ½" ID; 10/PK
  • Environmentally Preferable:
  • Orifice dimensions:
    8,73 mm (0,34375")
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Pk:
    10
  • Fitting material:
    PVDF
Frequently Bought Together