Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ProPlus PhyTip® columns (200 µL volume) containing 5 µL affinity media per column, Perkin Elmer. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTP-92-05-07
Supplier:
BIOTAGE
Expand image
ProPlus PhyTip® columns (200 µL volume) containing 5 µL affinity media per column, Perkin Elmer. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTP-92-05-07
Supplier:
BIOTAGE
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Cryovials
Supplier: VWR Collection
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Sample tubes
Supplier: Brand
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes with Socket Screw Caps
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Microcentrifuge tubes, 2,0 ml
Supplier: Brand
Multiple well cell culture plates, Costar®
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Cell culture flasks
Supplier: TPP
Serological pipettes
Supplier: Greiner Bio-One