0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ProPlus LX PhyTip® columns (200 µL volume) containing 20 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTB-92-20-26
Supplier:
BIOTAGE
Expand image
ProPlus LX PhyTip® columns (200 µL volume) containing 20 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTB-92-20-26
Supplier:
BIOTAGE
Frequently Bought Together
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF