Reversed phase PhyTip® columns (200 µL volume) containing 20 µL C18 (90 Å) silica per column, Beckman 1 * 96 items
Catalog # BITAPTB-92-20-18
Supplier: BIOTAGE
Reversed phase PhyTip® columns (200 µL volume) containing 20 µL C18 (90 Å) silica per column, Beckman 1 * 96 items
Catalog # BITAPTB-92-20-18
Supplier: BIOTAGE
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Test Tubes without Rim
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Polyester Films for ELISA and General Incubation
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR® Cryoboxes and Dividers
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF