0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
IMAC PhyTip® columns (200 µL volume) containing 20 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTB-92-20-03
Supplier:
BIOTAGE
Expand image
IMAC PhyTip® columns (200 µL volume) containing 20 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTB-92-20-03
Supplier:
BIOTAGE
Frequently Bought Together
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Sample Spoons, Disposable, SteriWare®
Supplier: Sampling Systems
VWR®, Test Tubes without Rim
Supplier: VWR Collection
Cell scrapers and cell lifters
Supplier: VWR Collection
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Centrifuge tubes, sterile, polypropylene, Corning®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Eppendorf LoBind® twin.tec® PCR Plates
Supplier: EPPENDORF
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF