SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
ProPlus LX PhyTip® columns (200 µL volume) containing 5 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
BITAPTB-92-05-26
:
BIOTAGE
ProPlus LX PhyTip® columns (200 µL volume) containing 5 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
BITAPTB-92-05-26
:
BIOTAGE
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR®, Cryoboxes with Dividers
: VWR Collection
VWR®, Stoppers, PE
: VWR Collection
VWR®, Straw Pipettes
: VWR Collection
VWR® Blender Bags
: VWR Collection
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Aluminium Foil Seals for PCR and Storage (96- and 384-Well Plates)
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
Petri dishes
: Greiner Bio-One
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF