ContactUsLinkComponent
Ion exchange PhyTip® columns (1 mL volume) containing 80 µL strong cation exchange resin per column, Beckman 1 * 96 items
Catalog # BITAPTB-91-80-13
Supplier:  BIOTAGE
Ion exchange PhyTip® columns (1 mL volume) containing 80 µL strong cation exchange resin per column, Beckman 1 * 96 items
Catalog # BITAPTB-91-80-13
Supplier:  BIOTAGE
Frequently Bought Together