ContactUsLinkComponent
ProPlus LX PhyTip® columns (1 mL volume) containing 40 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTB-91-40-26
Supplier:  BIOTAGE
ProPlus LX PhyTip® columns (1 mL volume) containing 40 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTB-91-40-26
Supplier:  BIOTAGE
Frequently Bought Together