Ion exchange PhyTip® columns (1 mL volume) containing 40 µL strong anion exchange resin per column, Beckman 1 * 96 items
Catalog # BITAPTB-91-40-11
Ion exchange PhyTip® columns (1 mL volume) containing 40 µL strong anion exchange resin per column, Beckman 1 * 96 items
Catalog # BITAPTB-91-40-11
Frequently Bought Together