Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Protein G PhyTip® columns (1 mL volume) containing 20 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTB-91-20-02
Supplier:
BIOTAGE
Expand image
Protein G PhyTip® columns (1 mL volume) containing 20 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
Catalog # BITAPTB-91-20-02
Supplier:
BIOTAGE
Frequently Bought Together
96-Well Polypropylene Storage Blocks, Corning®
Supplier: Corning
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR® Cell Culture Dishes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Pasteur pipettes, disposable
Supplier: Brand
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Deep well plates and cover mats
Supplier: Brand
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Eppendorf twin.tec® PCR Plates 96-well, 150 µl
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF