SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
IMAC PhyTip® columns (1 mL volume) containing 10 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
BITAPTB-91-10-03
:
BIOTAGE
IMAC PhyTip® columns (1 mL volume) containing 10 µL affinity media per column, Beckman. Incl. buffer kit. 1 * 96 items
BITAPTB-91-10-03
:
BIOTAGE
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR®, Transfer Pipettes, Graduated
: VWR Collection
J.T.Baker®, Plate 96
: Avantor
Transfer pipettes
: RATIOLAB
Cryoboxes, B69/B50/B80/B99-Series
: GLW
Colour coders for cryovials, CryoColorCode, Nunc™
: Thermo Fisher Scientific
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
: VWR Collection
J.T.Baker®, Plate Medio
: Avantor
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF