Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Buffer kit for 96 x 40 µL IMAC columns: Buffer E (125 mL), Buffer F (125 mL), and Buffer H (60 mL) 1 * 5 items
Catalog # BITABUF-91-40-03
Supplier:
BIOTAGE
Expand image
Buffer kit for 96 x 40 µL IMAC columns: Buffer E (125 mL), Buffer F (125 mL), and Buffer H (60 mL) 1 * 5 items
Catalog # BITABUF-91-40-03
Supplier:
BIOTAGE
Frequently Bought Together
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Transfer pipettes
Supplier: RATIOLAB
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Stoppers, PE
Supplier: VWR Collection
VWR®, Straw Pipettes
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF