READIUSE 10% TRITON X-100 *HYDROGEN PEROXIDE-AND CARBONYL-FREE* 1 * 100 mL
Catalog # MSPP60012
Supplier: MARKET SOURCE PART PROCESS
READIUSE 10% TRITON X-100 *HYDROGEN PEROXIDE-AND CARBONYL-FREE* 1 * 100 mL
Catalog # MSPP60012
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Pyrogallol for synthesis, Sigma-Aldrich®
Supplier: Merck
1-Butylboronic acid, Supelco®
Supplier: Merck
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
Supplier: PanReac AppliChem
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Xylene - Mixture of isomeres (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific