AMATEK FMOC-3-FLUORO-N-METHYL-L-PHENYLALANINE 1 * 20 g
Katalognr. MSPPA-8739-20G
Anbieter: MARKET SOURCE PART PROCESS
AMATEK FMOC-3-FLUORO-N-METHYL-L-PHENYLALANINE 1 * 20 g
Katalognr. MSPPA-8739-20G
Anbieter: MARKET SOURCE PART PROCESS
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Anbieter: Merck
HFIPA LC-MS 1 * 100 mL
Anbieter: Novartis
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Anbieter: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Anbieter: Merck
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Anbieter: Merck
1-Butylboronic acid, Supelco®
Anbieter: Merck
Ethylenediamine 99%
Anbieter: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
Anbieter: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Anbieter: Thermo Fisher Scientific
Ethylenediamine 99%
Anbieter: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Anbieter: Honeywell Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Anbieter: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
Anbieter: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
Anbieter: Honeywell Chemicals