FAMCICLOVIR RELATED COMPOUND F 1 * 50 mg
PHR2861-50MG
: Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
pH buffer solutions
: Mettler - Toledo