ESTRADIOL CYPIONATE 1 * 500 mg
Catalog # PHR2839-500MG
Supplier: Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW