PROCAINE N-OXIDE 1 * 100 mg
PHR2253-100MG
: Merck
Frequently Bought Together
BUFFER PH 9,21 DOSING BOTTLE 1 * 500 mL
: HAMILTON BONADUZ
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
: HAMILTON BONADUZ
pH buffers, DuraCal
: HAMILTON BONADUZ
CARNOSIC ACID 1 * 20 mg
: Merck
PCB-Mix 41 10 ng/µl 1 * 1 mL
: EHRENSTORFER
VWR® ARISTAR®, Molybdenum Standard Solutions for ICP-MS
: VWR Chemicals
URACIL ARABINOSIDE 1 * 100 mg
: Merck
VWR®, Flow Cup viscosity standards
: VWR Chemicals