AMLODIPINE RELATED COMPOUND K 1 * 50 mg
Catalog # PHR2056-50MG
Supplier: Merck
Frequently Bought Together
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Buffer solutions
Supplier: REAGECON
Buffer solutions, colour coded, Fluka™
Supplier: Honeywell Chemicals
1,2,3-Trimethylbenzene
Supplier: EHRENSTORFER
1H,1H,2H,2H-PERFLUOROHEXANESULFONIC ACID 100 µG/ML IN METHANOL:WATER 1 * 1 mL
Supplier: EHRENSTORFER
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, Oleanolic Acid
Supplier: Merck
Fluxametamide 1 * 25 mg
Supplier: EHRENSTORFER
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Potassium hydrogen phthalate 0.2 mol/l in aqueous solution Reag. Ph. Eur. 1070001
Supplier: VWR Chemicals
Conductivity standards, Potassium chloride, IUPAC
Supplier: Hach
Accessories for ion selective meters, LAQUAtwin
Supplier: HORIBA
pH Standard Solutions, IUPAC, Certified
Supplier: Hach