ContactUsLinkComponent
Masterflex® Fitting, PVDF Body with Polypropylene Insert, Female Luer Lock x Three-Way Stopcock
Masterflex® Fitting, PVDF Body with Polypropylene Insert, Female Luer Lock x Three-Way Stopcock
Catalog # MFLX40619-99
undefined
Masterflex® Fitting, PVDF Body with Polypropylene Insert, Female Luer Lock x Three-Way Stopcock
Catalog # MFLX40619-99
About this item
Build leakproof fluid handling systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
These one-way stopcocks feature an all-black PVDF construction.
  • Flow pattern:
    Three-Way
  • Description:
    Fitting, PVDF Body with Polypropylene Insert, Female Luer Lock x Three-Way Stopcock
  • Seq:
    06
  • Environmentally Preferable:
  • Wetted materials:
    Natural PVDF, Polypropylene
  • Pk:
    1
  • Inlet port:
    Female Luer Lock
  • Outlet port:
    Female Luer Lock
Frequently Bought Together