Orders LinkContactUsLinkComponent
Maybridge Screening library
Kit contents: Hand-selected 96 well plate, 50
Catalog # MAYLHTSPLATE
Maybridge Screening library
Catalog # MAYLHTSPLATE
CAS Number:  
Maybridge Screening library
Maybridge Screening library
Kit contents: Hand-selected 96 well plate, 50
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 50 ST
Product Details & Documents
The Maybridge portfolio consists of high-quality hit-like, lead-like and drug-like compounds which can generate valuable data in screening programs. They are developed specifically to support the pharmaceutical drug discovery process.
  • High-throughput screening for drug discovery
  • Maybridge hit-to-lead building blocks
  • Maybridge fragment libraries
See our dedicated Maybridge product page for more information and access to full library files, https://uk.vwr.com/cms/maybridge.
 : 96-well plates may be provided at mass ≥2 µmol or ≥0,5 mg as powder, dry films, or frozen DMSO solutions. 96- or 384-well plates may be provided at mass 0,1 µmol or 0,01 mg as dry films or frozen DMSO solutions. Compounds may also be ordered in vials at mass 2 µmol or 0,5 mg as powder or DMSO solutions.
We can also provide most major brands of plates and vials and we are happy to use those supplied by customers on request.
Specifications
Specifications
  • Pk:
    50
  • Description:
    Kit contents: Hand-selected 96 well plate