MIKROZID AF WIPES JUMBO REFILL (220PCS) 1 * 12 items
SCUE70003094
Frequently Bought Together
n-Heptane
: Merck
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
: PanReac AppliChem
Glycerine (synthetic) 99+%, pure
: Thermo Fisher Scientific
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Chloroform ≥99.9% stabilised, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
: PanReac AppliChem
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Toluene ≥99.3%, Laboratory Reagent
: Honeywell Chemicals
1-Pentanol 99+% ACS
: Thermo Fisher Scientific