ContactUsLinkComponent
TThe Ultralabel PRO-600 series is a desktop cleanroom label printer designed for critical environement applications. The printer offers enhanced functionality including print resolution of 600 dpi and a userfriendly TFT display 1 * 1 items
Catalog # UTAPULP600D
Supplier:  ULTRATAPE
TThe Ultralabel PRO-600 series is a desktop cleanroom label printer designed for critical environement applications. The printer offers enhanced functionality including print resolution of 600 dpi and a userfriendly TFT display 1 * 1 items
Catalog # UTAPULP600D
Supplier:  ULTRATAPE
Frequently Bought Together