Orders LinkContactUsLinkComponent
CytoFLEX SRT Software Upgrades
Activation key to upgrade the CytoFLEX SRT V0-B2-Y0-R3 Cell Sorter with additional parameters. Upon installation the instrument will have 3 active lasers (405 nm, 488 nm, 638 nm) and 10 fluorescence detectors in the V5-B2-Y0-R3 configuration
  BECMC65146
CytoFLEX SRT Software Upgrades
  BECMC65146
CytoFLEX SRT Software Upgrades
Activation key to upgrade the CytoFLEX SRT V0-B2-Y0-R3 Cell Sorter with additional parameters. Upon installation the instrument will have 3 active lasers (405 nm, 488 nm, 638 nm) and 10 fluorescence detectors in the V5-B2-Y0-R3 configuration
1 * 1 ST
  • Description:
    Activation key to upgrade the CytoFLEX SRT V0-B2-Y0-R3 Cell Sorter with additional parameters. Upon installation the instrument will have 3 active lasers (405 nm, 488 nm, 638 nm) and 10 fluorescence detectors in the V5-B2-Y0-R3 configuration
  • Seq:
    10