SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
TUBING L/FLAT 12X500FT 3CORE 0.004MM 1 * 1 Roll
FISHSTCK0057-002
:
FISHER CONTAINER
TUBING L/FLAT 12X500FT 3CORE 0.004MM 1 * 1 Roll
FISHSTCK0057-002
:
FISHER CONTAINER
Frequently Bought Together
VWR® Nitrile Light Long, Nitrile Gloves
: VWR Collection
Neoprene gloves, Microflex® NeoTouch® 25-101 / 25-201
: Ansell
Shoe covers, PP/CPE
: HUBEI ORIENT INTL TRADING
VWR®, Nitrile Gloves
: VWR Collection
Cleanroom gloves, polychloroprene, DermaShield® 73-711 / 73-721
: Ansell
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Labels, Hazardous Substance, GHS Labels
: VWR Collection
Cleanroom gloves, polychloroprene, BioClean Fusion™
: Nitritex
VWR® Cleanroom Gloves, Nitrile, Sterile, ISO 5, 400 mm
: VWR Collection
Overalls, 4500
: 3M
VWR®, Latex Gloves
: VWR Collection