ContactUsLinkComponent
Masterflex® Fitting, Natual Polypropylene, Tee, Hose Barb to Threaded Adpater, 3/8" ID x 3/8" ID x 1/4" NPT(M); 100/PK
Masterflex® Fitting, Natual Polypropylene, Tee, Hose Barb to Threaded Adpater, 3/8" ID x 3/8" ID x 1/4" NPT(M); 100/PK
Catalog # MFLX40640-75
CE certificate
undefined
Masterflex® Fitting, Natual Polypropylene, Tee, Hose Barb to Threaded Adpater, 3/8" ID x 3/8" ID x 1/4" NPT(M); 100/PK
Catalog # MFLX40640-75
CE certificate
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/8" ID x 3/8" ID x 1/4" NPT(M)
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
  • Barb size/tubing int.Ø (port 2):
    ⅜"
  • Connection Type:
    Thread to Barb
  • Flow pattern:
    T
  • Description:
    Fitting, Natual Polypropylene, Tee, Hose Barb to Threaded Adpater, ⅜" ID × ⅜" ID × ¼" NPT(M); 100/PK
  • Seq:
    17
  • Environmentally Preferable:
  • Model:
    40640-75
  • Orifice dimensions:
    0,125"
  • Sterilisation:
    Ethylene oxide (ETO) or gamma
  • Warranty:
    Out of box
  • Pk:
    100
  • Fitting material:
    Polypropylene
Frequently Bought Together